missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) Chloride, Ultra Dry, 99.9985% (Metal Basis), Alfa Aesar™

$70.35 - $70.35

Chemical Identifiers

CAS 7772-99-8
Physical Form Powder
Assay Percent Range 99.9%, metal basis
Molecular Formula SnCl2
Formula Weight 189.62
MDL Number MFCD00011241
Synonym Stannous chloride
Chemical Name or Material Tin(II) Chloride, Ampouled under argon
Grade Ultra dry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3572303 SDS Alfa Aesar
3572303
1g Each for $70.35
AA3572306 SDS Alfa Aesar
3572306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3572322 SDS Alfa Aesar
3572322
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3572314 SDS Alfa Aesar
3572314
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7772-99-8
99.9%, metal basis
189.62
Stannous chloride
Ultra dry
Powder
SnCl2
MFCD00011241
Tin(II) Chloride, Ampouled under argon

Specifications

7772-99-8
99.9%, metal basis
189.62
Stannous chloride
Air, moisture sensitive
Tin(II) Chloride, Ampouled under argon
Powder
SnCl2
MFCD00011241
3.95
1g
Ultra dry

Safety and Handling

Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Harmful if swallowed. Causes burns. May cause sensitization by skin contact.

DOTInformation : DOT Class: 8

EINECSNumber : 231-868-0