missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1089414

 View more versions of this product

Catalog No. AA1089414



Specifications

Specifications

Tin(II) chloride hydrate
(metals basis)
SnCl2·xH2O
UN3260
Stannous chloride hydrate
Cl[Sn]Cl
189.61
CHEBI:78067
Crystalline Powder
Puratronic™
Odorless
White
25g
7772-99-8
Cl2Sn
MFCD00011241
14,8783
AXZWODMDQAVCJE-UHFFFAOYSA-L
dichlorotin
24479
189.62 (Anhydrous)
99.995%
2.71g/mL
Air Sensitive
43°C to 46°C

Chemical Identifiers

7772-99-8
189.61
AXZWODMDQAVCJE-UHFFFAOYSA-L
24479
dichlorotin
Cl2Sn
MFCD00011241
Stannous chloride hydrate
CHEBI:78067
Cl[Sn]Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P280-P262-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : XP8850000

TSCA : TSCA

Documents