Learn More
Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011536A3
Description
Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(II) chloride dihydrate | |
Reagent Grade | |
Crystalline | |
SnCl2·2H2O | |
Stannous chloride dihydrate | |
225.65 (189.62 Anhydrous) | |
Air Sensitive |
43°C to 46°C | |
10025-69-1 | |
2 kg | |
MFCD00149863 | |
Soluble in dilute and concentrated HCl,alcohol,ethyl acetate,glacial acetic acid,and NaOH solution. Decomposes in excess water. | |
Reagent | |
2.71 g/mL |
Chemical Identifiers
10025-69-1 | |
SnCl2·2H2O | |
MFCD00149863 | |
Stannous chloride dihydrate |
Safety and Handling
P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
H290-H314-H317-H332-H335-H373
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 600-045-1
RTECSNumber : XP8850000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only