missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4440922

 View more versions of this product

Catalog No. AA4440922



Specifications

Specifications

Tin(II) chloride dihydrate
10025-69-1
SnCl2·2H2O
Soluble in dilute and concentrated hydrochloric acid,alcohol,ethyl acetate,glacial acetic acid,and sodium hydroxide solution. Decomposes in excess water.
Crystalline
2.71 g/mL
43°C to 46°C
For analysis ACS
98.0-103.0%
MFCD00149863
225.65 (189.62 Anhydrous)
ACS Reagent
Air Sensitive
100g

Chemical Identifiers

10025-69-1
SnCl2·2H2O
MFCD00149863
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H314-H317-H332-H335-H373

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 600-045-1

RTECSNumber : XP8850000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Documents