Learn More
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196800050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
652.0°C | |
37.0°C to 38.0°C | |
10025-69-1 | |
Adhering Crystalline Powder or Crystals | |
96% min. (Iodimetry) | |
SnCl2·2H2O | |
01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68 | |
Stannous chloride dihydrate | |
225.63 | |
97% | |
2.7100g/mL |
Tin(II) chloride dihydrate | |
97% | |
White or Colorless | |
5 g | |
Cl2Sn·2H2O | |
MFCD00149863 | |
15, 8910 | |
Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid | |
225.63 | |
Glass Bottle |
Chemical Identifiers
10025-69-1 | |
225.63 | |
Stannous chloride dihydrate |
Cl2Sn·2H2O | |
MFCD00149863 |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through prolon
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
RTECSNumber : XP8850000
RUO – Research Use Only