Please login to your online account to display your discounted pricing

Tin(II) acetate, 95%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tin(II) acetate
CAS 638-39-1
Physical Form Powder
Assay Percent Range 95%
Molecular Formula Sn(OOCCH3)2
Formula Weight 236.8
MDL Number MFCD00054408
Synonym Stannous acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2236106

  SDS
alfa aesar
2236106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2236114

  SDS
alfa aesar
2236114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.8
Physical Form Powder
Quantity 5g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
EINECS Number 211-335-9
Chemical Name or Material Tin(II) acetate
Health Hazard 1 H331-H302-H312: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin.
Appearance White powder
Assay Percent Range 95%
CAS 638-39-1
Synonym Stannous acetate
MDL Number MFCD00054408
Molecular Formula Sn(OOCCH3)2
Safety and Handling

H331-H302-H312: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin.