missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) 2-ethylhexanoate, tech.

Manufacturer:  Alfa Aesar™ 1859036

 View more versions of this product

Catalog No. AA1859036



Specifications

Specifications

Tin(II) 2-ethylhexanoate
500g
1.251g/mL
1.4940
MFCD00002676
KSBAEPSJVUENNK-UHFFFAOYSA-L
2-ethylhexanoate;tin(2+)
9318
Liquid
301-10-0
>110° (230°F)
Colorless to Yellow
C16H30O4Sn
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
405.122
405.11
Technical

Chemical Identifiers

301-10-0
405.122
KSBAEPSJVUENNK-UHFFFAOYSA-L
9318
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
C16H30O4Sn
MFCD00002676
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
2-ethylhexanoate;tin(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MO7870000

TSCA : TSCA

Documents