Please login to your online account to display your discounted pricing

Tin ICP Standard, 1mL = 1mg Sn (1,000ppm Sn) in 3% HNO3, Ricca Chemical

$141.41 - $141.41

Chemical Identifiers

Chemical Name or Material Tin ICP Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.89
CAS Max % 5.09
CAS 7664-39-3
CAS Min % 0.12
CAS Max % 0.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PSN1KN-100

 
ricca chemical company
RIPSN1KN100
Poly natural 100mL Each for $141.41

PSN1KN-500

 
ricca chemical company
RIPSN1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
Concentration or Composition Notes 1,000ppm Sn
Equivalence 1 mL = 1 mg Sn
Chemical Name or Material Tin ICP Standard
CAS Max % 0.12
CAS 7440-31-5
CAS Min % 0.09
CAS Max % 0.1
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
Packaging Poly natural
CAS Min % 4.89
CAS Max % 5.09
CAS 7664-39-3
CAS Min % 0.12