missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin ICP Standard, 1mL = 1mg Sn (1,000ppm Sn) in 3% HCl, Ricca Chemical

$183.41 - $183.41

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PSN1KH-100
View Documents
Ricca Chemical Company
RIPSN1KH100
100mL Poly natural Each for $183.41
 
PSN1KH-500
View Documents
Ricca Chemical Company
RIPSN1KH500
500mL Poly natural N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7440-31-5
118.71
5352426
tin
Sn
ATJFFYVFTNAWJD-UHFFFAOYSA-N
CHEBI:32990
[Sn]

Specifications

7440-31-5,7647-01-0,7732-18-5
86.0,15.48,0.09
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
Liquid
1,000ppm Sn
Colorless
Tin ICP Standard
83.0,14.87,0.08
Sn
[Sn]
118.71
CHEBI:32990
Poly natural
1 mL = 1 mg Sn
100mL