missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tin ICP Standard, 10,000 ppm Sn in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PSN10KN100
Specifications
Tin ICP Standard | |
Natural LDPE Bottle | |
84.0,12.56,1.04,0.84 | |
Colorless | |
Sn | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
1 mL = 10 mg Sn |
7440-31-5,7697-37-2,7664-39-3,7732-18-5 | |
100 mL | |
87.0,13.07,1.09,0.87 | |
10,000ppm Sn | |
MFCD00133862 | |
[Sn] | |
118.71 | |
CHEBI:32990 |
Chemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
CHEBI:32990 | |
[Sn] |
Sn | |
MFCD00133862 | |
5352426 | |
tin |