Please login to your online account to display your discounted pricing

Tin ICP Standard, 1mL = 10mg Sn (10,000ppm Sn) in 3% HNO3, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Tin ICP Standard
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 87.0
CAS 7697-37-2
CAS Min % 12.56
CAS Max % 13.07
CAS 7664-39-3
CAS Min % 1.04
CAS Max % 1.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PSN10KN-100

 
ricca chemical company
RIPSN10KN100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PSN10KN-50

 
ricca chemical company
RIPSN10KN50
Poly natural 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Concentration or Composition Notes 10,000ppm Sn
Chemical Name or Material Tin ICP Standard
CAS 7732-18-5
CAS Min % 84.0
CAS Min % 12.56
CAS 7664-39-3
Equivalence 1 mL = 10 mg Sn
CAS Max % 1.09
CAS 7440-31-5
CAS Min % 0.84
CAS Max % 0.87
CAS Max % 87.0
CAS 7697-37-2
Packaging Poly natural
CAS Max % 13.07
CAS Min % 1.04