missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin foil, 2.0mm (0.08 in.) thick, Puratronic™, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 11456GH

 View more versions of this product

Catalog No. AA11456GH



Specifications

Specifications

Tin foil
100 x 100mm
7440-31-5
Sn
flake
[Sn]
Odorless
5352426
Foil
Puratronic™
≈9.1g/25x25mm
2.0mm (0.08 in.) thick
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.9985%

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents