missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin Foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.9985% (Metals basis), Alfa Aesar™

Form: Approx. 9.1g/25 x 50mm

Manufacturer:  Alfa Aesar 11457HL

 View more versions of this product

Catalog No. AA11457HL



Specifications

Specifications

Tin
7440-31-5
99.9985% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Silver to Gray
100 x 250mm
Silver Gray foil
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
Odorless
5352426
Foil, 1.0mm (0.04 in.) thick

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]
Documents