missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin foil, 0.5mm (0.02 in.) thick, Puratronic™, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 11458FI

 View more versions of this product

Catalog No. AA11458FI



Specifications

Specifications

Tinfoil
50 x 50mm
7440-31-5
Sn
flake, metallic, powder, silver matt powder, stannum, tin, elemental, tin, metal, wang, zinn, zinn german
[Sn]
Odorless
5352426
Foil
Puratronic™
≈9.1g/50x50mm
0.5mm (0.02 in.) thick
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.9985%

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake, metallic, powder, silver matt powder, stannum, tin, elemental, tin, metal, wang, zinn, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

EINECSNumber : 231-141-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents