missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin Foil, 0.5mm (0.02 in.) thick, 99.8% (Metals basis), Alfa Aesar™

Form: Approx. 9.1g/50 x 50mm

Manufacturer:  Alfa Aesar 43234GH

 View more versions of this product

Catalog No. AA43234GH



Specifications

Specifications

Tin
7440-31-5
7.31g/cm3
Sn
flake
[Sn]
Odorless
5352426
Foil, 0.5mm (0.02 in.) thick
100 x 100mm
Silver Gray foil
99.8% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Silver to Gray

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Documents