missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin foil, 0.33mm (0.013 in.) thick, 99+% (metals basis)

Manufacturer:  Alfa Aesar™ 14636RE

 View more versions of this product

Catalog No. AA14636RE



Specifications

Specifications

Tin foil
5 x 150 x 270mm
7440-31-5
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
Odorless
tin
5352426
Foil
≈100g/15x27cm
0.33mm (0.013 in.) thick
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
[Sn]
118.71
CHEBI:32990
≥99%

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents