missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin foil, 0.18mm (0.007 in.) thick, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 43308FI

 View more versions of this product

Catalog No. AA43308FI



Specifications

Specifications

Tin foil
50 x 50mm
7440-31-5
Sn
flake
[Sn]
Odorless
5352426
Foil
≈3.3g/50x50mm
0.18mm (0.007 in.) thick
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.9%

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents