Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin foil, 0.05mm (0.002 in.) thick, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Tin foil
CAS 7440-31-5
Name Note 0.05mm (0.002 in.) thick
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38538FY

 
alfa aesar™
38538FY
50x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tin foil
TSCA TSCA
Name Note 0.05mm (0.002 in.) thick
Physical Form Foil
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00133862
CAS 7440-31-5
Odor Odorless
Weight ≈1.8g/50x100mm
Quantity 50x100mm