missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin, AAS standard solution, Sn 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 88112AE

 View more versions of this product

Catalog No. AA88112AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tin
7440-31-5
Sn 1000;mug/mL
MFCD00133862
[Sn]
118.71
CHEBI:32990
Characteristic
Approx. 1g/cm3 at 20°C
AAS standard solution, Sn 1000μg/mL
Liquid
Sn
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
Liquid
23 hPa (17mm Hg) at 20°C
100mL

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
CHEBI:32990
[Sn]
Sn
MFCD00133862
5352426
tin
Safety and Handling

Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-595-7

SDS Documents