Please login to your online account to display your discounted pricing

Tin, AAS standard solution, Sn 1000μg/mL, Alfa Aesar™, Specpure™

Chemical Identifiers

Chemical Name or Material Tin
Name Note AAS standard solution, Sn 1000μg/mL
Physical Form Liquid
Assay Percent Range Sn 1000;mug/mL
Molecular Formula Sn in 20% HCl
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88112AE

  SDS
alfa aesar
88112AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA88112AP

  SDS
alfa aesar
88112AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100mL
DOT Information DOT Class: 8, Packaging Group: II
Physical Form Liquid
EINECS Number 231-595-7
Density Approx. 1g/cm3 at 20°C
Assay Percent Range Sn 1000;mug/mL
Name Note AAS standard solution, Sn 1000μg/mL
MDL Number MFCD00133862
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Appearance Liquid
Odor Characteristic
Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
Chemical Name or Material Tin
Molecular Formula Sn in 20% HCl
Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.