Learn More
Tin, 99.8%, powder, -325 mesh, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 317805000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Gray | |
-325 mesh | |
7440-31-5 | |
7.3000g/mL | |
Sn | |
MFCD00133862 | |
01,1168; 02,414; 11,519; 12,486; 13,298; 17,333 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
118.69 | |
Plastic bottle |
Tin | |
231.9°C | |
2270.0°C | |
99.8% | |
Sn | |
500 g | |
15, 9603 | |
Solubility in water: insoluble. Other solubilities: soluble in acids and hot potassium hydroxide, solutions | |
[Sn] | |
118.71 | |
CHEBI:32990 | |
99.8% | |
Powder |
Chemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
5352426 | |
tin |
Sn | |
MFCD00133862 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
CHEBI:32990 | |
[Sn] |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only