Please login to your online account to display your discounted pricing

Tin, 99.5%, powder, -100 mesh, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC317815000

 View more versions of this product

Catalog No. AC317815000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 118.69
Quantity 500g
Color Gray
Density 7.3000g/mL
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Merck Index 15, 9603
Packaging Plastic bottle
CAS 7440-31-5
Total Trace Metal Impurities 0.5% max.
Name Note -100 mesh
Assay Percent Range 99.5%
Chemical Name or Material Tin
Molecular Formula Sn
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula Sn
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00133862

Specifications

Chemical Name or Material Tin
Name Note -100 mesh
CAS 7440-31-5
Physical Form Powder
Assay Percent Range 99.5%
Molecular Formula Sn
Linear Formula Sn
Formula Weight 118.69
MDL Number MFCD00133862
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents