missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin, 30 mesh, Reagent ACS, Granular, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 420995000

 View more versions of this product

Catalog No. AC420995000


 

Specifications Description & Specifications

Specifications

≥99.5%
Gray
ACS Reagent Grade
15, 9603
01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
50ppm max.
Sn
Tin
200ppm max.
100ppm max.
118.69
Metallic Granular Powder
500g
7.3000g/mL
Sn
7440-31-5
MFCD00133862
50ppm max.
Glass bottle
1ppm max.

Chemical Identifiers

7440-31-5
Sn
MFCD00133862
≥99.5%
Sn
118.69
Tin
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-141-8

RTECSNumber : XP7320000

TSCA : TSCA