missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TIMP-2 Ab-5, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ TIMP-2 Ab-5, Mouse Monoclonal Antibody.

$337.55 - $1,972.04

Specifications

Antigen TIMP-2 Ab-5
Clone 3A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 9
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1485B View Documents Thermo Scientific™
MS1485B
Biotinylated Ab with BSA and Azide 1mL Each for $1,972.04
 
MS1485B0 View Documents Thermo Scientific™
MS1485B0
Biotinylated Ab with BSA and Azide 100μL Each for $451.73
 
MS1485B1 View Documents Thermo Scientific™
MS1485B1
Biotinylated Ab with BSA and Azide 500μL Each for $1,230.33
 
MS1485P View Documents Thermo Scientific™
MS1485P
Purified Ab with BSA and Azide 1mL Each for $1,580.59
 
MS1485P0 View Documents Thermo Scientific™
MS1485P0
Purified Ab with BSA and Azide 100μL Each for $337.55
 
MS1485P1 View Documents Thermo Scientific™
MS1485P1
Purified Ab with BSA and Azide 500μL Each for $949.73
 
MS1485P1ABX View Documents Thermo Scientific™
MS1485P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $1,050.82
 
MS1485PABX View Documents Thermo Scientific™
MS1485PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,717.59
 
MS1485R7 View Documents Thermo Scientific™
MS1485R7
Ready-to-use for IHC (UVLP) 7mL Each for $787.88
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ TIMP-2 Ab-5, Mouse Monoclonal Antibody.

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Host Species: Mouse

Clone: 3A4

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein

Molecular Weight: 21kDa

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TIMP-2 Ab-5
Immunohistochemistry (Paraffin), Western Blot
Murine
Proteases And Inhibitors
A synthetic peptide from the N-terminal region of human TIMP-2 protein
3A4
Monoclonal
RUO
Primary
IgG2a kappa