missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TIMP-1 Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ TIMP-1 Ab-2, Mouse Monoclonal Antibody.

$342.63 - $1,978.37

Specifications

Antigen TIMP-1 Ab-2
Clone 102D1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 8
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS608B View Documents Thermo Scientific™
MS608B
Biotinylated Ab with BSA and Azide 1mL Each for $1,978.37
 
MS608B0 View Documents Thermo Scientific™
MS608B0
Biotinylated Ab with BSA and Azide 100μL Each for $453.76
 
MS608B1 View Documents Thermo Scientific™
MS608B1
Biotinylated Ab with BSA and Azide 500μL Each for $1,247.86
 
MS608P View Documents Thermo Scientific™
MS608P
Purified Ab with BSA and Azide 1mL Each for $1,593.18
 
MS608P0 View Documents Thermo Scientific™
MS608P0
Purified Ab with BSA and Azide 100μL Each for $342.63
 
MS608P1 View Documents Thermo Scientific™
MS608P1
Purified Ab with BSA and Azide 500μL Each for $953.00
 
MS608P1ABX View Documents Thermo Scientific™
MS608P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $1,037.00
 
MS608R7 View Documents Thermo Scientific™
MS608R7
Ready-to-use for IHC (UVLP) 7mL Each for $1,018.51
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ TIMP-1 Ab-2, Mouse Monoclonal Antibody.

Tissue inhibitor of metalloproteinases-1 (TIMP-1) and Tissue inhibitor of metalloproteinases-2 (TIMP-2) are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-1 forms a complex of 1:1 stoichiometry with activated interstitial collagenase, activated stromelysin, active form of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A), and latent and active forms 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMPs inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Host Species: Mouse

Clone: 102D1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human TIMP-1

Molecular Weight: 28.5kDa

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fetal lung cell line HFL-1; Bladder, breast and ovarian CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TIMP-1 Ab-2
Immunohistochemistry (Paraffin), Western Blot
Murine
Proteases And Inhibitors
Recombinant human TIMP-1
102D1
Monoclonal
RUO
Primary
IgG1