Please login to your online account to display your discounted pricing

TIM4 Rat anti-Mouse, PerCP-eFluor 710, Clone: 54 (RMT4-54), eBioscience™

Rat Monoclonal Antibody

$187.05 - $477.05

Specifications

Antigen TIM4
Clone 54 (RMT4-54)
Host Species Rat
Gene Alias TIMD4, SMUCKLER
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123845

 
life technologies
LS46586680
25μg Each for $187.05

501123846

 
life technologies
LS46586682
100μg Each for $477.05
Description & Specifications

Specifications

Antigen TIM4
Clone 54 (RMT4-54)
Host Species Rat
Gene Alias TIMD4, SMUCKLER
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody 54 (RMT4-54) recognizes mouse Tim-4 (T cell, Ig, mucin). Unlike other Tim family members, Tim-4 is not expressed on T lymphocytes but rather found on antigen presenting cells such as lymphoid and myeloid DC’s and macrophages. Tim-4 has been shown to interact with Tim-1 thereby suggesting a role of Tim-4 in the balance of Th1/Th2 regulation. Additionally Tim-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. Expression is found on a subset of CD11c+ splenic cells.