missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TIM4 Rat anti-Mouse, PE, Clone: 54 (RMT4-54), eBioscience™

Rat Monoclonal Antibody

$143.88 - $394.68

Specifications

Antigen TIM4
Clone 54 (RMT4-54)
Host Species Rat
Gene Alias TIMD4, SMUCKLER
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010912 Life Technologies
LS12586680
25μg Each for $143.88
5010913 Life Technologies
LS12586682
100μg Each for $394.68
Specifications

Description

The monoclonal antibody 54 recognizes mouse Tim-4 (T cell, Ig, mucin). Unlike other Tim family members, Tim-4 is not expressed on T lymphocytes but rather found on antigen presenting cells such as lymphoid and myeloid DC’s and macrophages. Tim-4 has been shown to interact with Tim-1 thereby suggesting a role of Tim-4 in the balance of Th1/Th2 regulation. Additionally Tim-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. Expression is found on a subset of CD11c+ splenic cells.
Description & Specifications

Specifications

TIM4
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
54 (RMT4-54)
TIMD4, SMUCKLER
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL