missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tilorone Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America T28361G
Specifications
Tilorone Dihydrochloride | |
234°C | |
MFCD00134071 | |
2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride | |
[H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1 | |
483.47 | |
483.47 | |
Crystalline Powder |
27591-69-1 | |
C25H36Cl2N2O3 | |
1 g | |
BSVYJQAWONIOOU-UHFFFAOYSA-N | |
dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride | |
33958 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
27591-69-1 | |
483.47 | |
BSVYJQAWONIOOU-UHFFFAOYSA-N | |
33958 | |
[H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1 |
C25H36Cl2N2O3 | |
MFCD00134071 | |
2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride | |
dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride |
Safety and Handling
RTECSNumber : LL9000000
TSCA : No