missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TIGIT, PerCP-eFluor™ 710, clone: GIGD7, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS46950182

 View more versions of this product

Catalog No. 501128768



Description

Description

Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.

PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications

Specifications

TIGIT
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P86176
VSIG9, VSTM3, WUCAM, V-set and immunoglobulin domain containing 9, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain containing 3, V-set and transmembrane domain-containing protein 3, Washington University cell adhesion molecule
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
GIGD7
PerCP-eFluor™ 710
Tigit
Liquid
Tigit
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
100043314
Documents

For Research Use Only.