missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TIGIT Rat anti-Mouse, PerCP-eFluor 710, Clone: GIGD7, eBioscience™

Rat Monoclonal Antibody

$191.40 - $487.20

Specifications

Antigen TIGIT
Clone GIGD7
Host Species Rat
Gene Alias VSTM3, WUCAM
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123954 Life Technologies
LS46950180
25μg Each for $191.40
501128768 Life Technologies
LS46950182
100μg Each for $487.20
Specifications

Description

The GIGD7 monoclonal antibody reacts with mouse T cell immunoreceptor with Ig and ITIM domains (TIGIT, VSTM3, WUCAM), a 26kDa protein that is a member of the poliovirus receptor (PVR) family. In mice, the expression of TIGIT has been reported in follicular T helper cells, while in humans, the expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells. TIGIT is not expressed on B cells or naïve CD4+ T cells. TIGIT is upregulated on CD4+ T cells following activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK cells and T cells with antigen presenting cells, fibroblasts and endothelial cells that express PVR and PVR-like proteins.
Description & Specifications

Specifications

TIGIT
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
GIGD7
VSTM3, WUCAM
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL