missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TIGIT Rat anti-Mouse, eFluor® 660, Clone: GIGD7, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS50950180

 View more versions of this product

Catalog No. 501128665



Description

Description

Description: The GIGD7 monoclonal antibody reacts with mouse T cell immunoreceptor with Ig and ITIM domains (TIGIT, VSTM3, WUCAM), a 26 kDa protein that is a member of the poliovirus receptor (PVR) family. In mice, the expression of TIGIT has been reported in follicular T helper cells, while in humans, the expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells. TIGIT is not expressed on B cells or naive CD4+ T cells. TIGIT is upregulated on CD4+ T cells following activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK cells and T cells with antigen presenting cells, fibroblasts and endothelial cells that express PVR and PVR-like proteins. Applications Reported: This GIGD7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This GIGD7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647.

eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications

Specifications

TIGIT
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P86176
VSIG9, VSTM3, WUCAM, V-set and immunoglobulin domain containing 9, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain containing 3, V-set and transmembrane domain-containing protein 3, Washington University cell adhesion molecule
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
GIGD7
eFluor® 660
Tigit
Liquid
Tigit
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
100043314
Documents

For Research Use Only.