missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TIGIT Mouse anti-Human, PerCP-eFluor 710, Clone: MBSA43, eBioscience™

Mouse Monoclonal Antibody

$200.64 - $496.32

Specifications

Antigen TIGIT
Clone MBSA43
Host Species Murine
Gene Alias VSTM3, WUCAM
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245942 Life Technologies
LS46950041
25 Tests Each for $200.64
50245943 Life Technologies
LS46950042
100 Tests Each for $496.32
Specifications

Description

The MBSA43 monoclonal antibody reacts with human T cell immunoreceptor with Ig and ITIM domains (TIGIT), which is also known as VSTM3 or WUCAM. This 26kDa protein is a member of the poliovirus receptor (PVR) family. The expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but it is not expressed on B cells or naïve CD4+ T cells. TIGIT may be upregulated on naïve CD4+ T cells upon activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK and T cells with antigen presenting cells, fibroblasts, and endothelial cells that express PVR and PVR-like proteins.

MBSA43 is reported to crossreact with Rhesus macaque.
Description & Specifications

Specifications

TIGIT
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MBSA43
VSTM3, WUCAM
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test