Please login to your online account to display your discounted pricing

TIGIT Mouse anti-Human, FITC, Clone: MBSA43, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $216.05

Specifications

Antigen TIGIT
Clone MBSA43
Host Species Mouse
Gene Alias VSTM3, WUCAM
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501129272

 
affymetrix
11950041
25 tests Each for $216.05
Description & Specifications

Specifications

Antigen TIGIT
Clone MBSA43
Host Species Mouse
Gene Alias VSTM3, WUCAM
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The MBSA43 monoclonal antibody reacts with human T cell immunoreceptor with Ig and ITIM domains (TIGIT), which is also known as VSTM3 or WUCAM. This 26kDa protein is a member of the poliovirus receptor (PVR) family. The expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but it is not expressed on B cells or naïve CD4+ T cells. TIGIT may be upregulated on naïve CD4+ T cells upon activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK and T cells with antigen presenting cells, fibroblasts, and endothelial cells that express PVR and PVR-like proteins.

MBSA43 is reported to crossreact with Rhesus macaque.