missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tid-1 Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tid-1 Ab-2, Mouse Monoclonal Antibody.

$267.63 - $1,375.45

Specifications

Antigen Tid-1 Ab-2
Clone RS13
Applications Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1564P Thermo Scientific™ Lab Vision™
MS1564P
200μg/mL 1mL Each for $1,252.22
MS1564P0 Thermo Scientific™ Lab Vision™
MS1564P0
200μg/mL 100μL Each for $267.63
MS1564P1ABX Thermo Scientific™ Lab Vision™
MS1564P1ABX
1mg/mL 100μL Each for $841.53
MS1564PABX Thermo Scientific™ Lab Vision™
MS1564PABX
1mg/mL 200μL Each for $1,375.45
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tid-1 Ab-2, Mouse Monoclonal Antibody.

TID1 is a human homolog of the Drosophila tumor suppressor lethal (2) tumerous imaginal discs, l(2)tid and encodes two mitochondrial matrix localized splice variants of human Tid-1 designated hTid-1S and hTid-1L. These proteins are the conserved members of the DnaJ family of proteins which act as cochaperons for mitochondrial Hsp70. They contain a conserved tetrahedrical J domain which binds to Hsp70 chaperones and activates their ATPase activity. Expression of hTid-1L increases apoptosis induced by DNA damaging agents as mitomycin-C and TNF-α. A J-domain mutant of hTid-1L can dominantly suppress apoptosis and in sharp contrast the J-domain mutant of hTid-1S increases apoptosis. Expression of hTid-1S and hTid-1L affects cytochrome c release from the mitochondria and caspase 3 activation, while activation of caspase 8 is unaffected. It is strongly suggested that these two splice variants exert their anti- and pro- apoptotic effects through discrete substrates and actiuvities. Hence the relative abundance of these proteins or their substrates may allow the mitochondria to dampen or enhance the apoptotic signals.

Host Species: Mouse

Clone: RS13

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Immunogen: Recombinant hTid-1 protein

Molecular Weight: 40 and 43kDa

Positive Control: HeLa and Jurkat cells

Cellular Localization: Mitochondria

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Tid-1 Ab-2
Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Unlabeled
RUO
Primary
RS13
Monoclonal
Murine
Apoptosis
Recombinant hTid-1 protein