Please login to your online account to display your discounted pricing

Alfa Aesar™ Ticarcillin disodium and clavulanate potassium (15:1), 50mg/mL in distilled water, sterile-filtered

$232.04 - $232.04

Chemical Identifiers

Chemical Name or Material Ticarcillin disodium and clavulanate potassium (15:1)
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD26967928
Synonym Timentin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ672958EQ

 
alfa aesar™
J672958EQ
5 x 1mL Pack for $232.04
Description & Specifications

Specifications

Chemical Name or Material Ticarcillin disodium and clavulanate potassium (15:1)
Name Note 50mg/mL in distilled water, sterile-filtered
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Physical Form Liquid
MDL Number MFCD26967928
Synonym Timentin
Quantity 5 x 1mL
Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

-30° to -10°C