missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tiaprofenic Acid, TCI America™

Manufacturer:  TCI America T3175100MG

 View more versions of this product

Catalog No. T3175100MG



Specifications

Specifications

Tiaprofenic Acid
99°C
33005-95-7
MFCD00866089
2-(5-Benzoylthiophen-2-yl)propionic Acid
CC(C1=CC=C(S1)C(=O)C2=CC=CC=C2)C(=O)O
260.307
CHEBI:32221
Crystal-Powder at 20°C
White-Yellow
100mg
C14H12O3S
2811
GUHPRPJDBZHYCJ-UHFFFAOYSA-N
2-(5-benzoylthiophen-2-yl)propanoic acid
5468
260.31
≥98.0% (GC,T)

Chemical Identifiers

33005-95-7
260.307
GUHPRPJDBZHYCJ-UHFFFAOYSA-N
5468
2-(5-benzoylthiophen-2-yl)propanoic acid
C14H12O3S
MFCD00866089
2-(5-Benzoylthiophen-2-yl)propionic Acid
CHEBI:32221
CC(C1=CC=C(S1)C(=O)C2=CC=CC=C2)C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : XM7580000

TSCA : No

Documents