man
Test
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
2017-Fisher-Scientific-Logo
2017-Fisher-Scientific-Logo
Sign In
Don't have an account?
Create an Account
Account
Main Menu
Account
Account Dashboard
Select Account
Products
Antibodies
Products
Antibodies
Antibody Panels and Kits
Antibody Production and Purification Reagents and Kits
Bovine Secondary Antibodies
Buffers and Standards
Donkey Secondary Antibodies
Goat Secondary Antibodies
Primary Antibodies for ELISA
Primary Antibodies for Immunohistochemistry
Primary Antibodies for Western Blotting
Secondary Antibodies
Chemicals
Products
Chemicals
Acids
Analytical Chromatography
Bases
Biochemical Reagents
Buffers and Standards
Organic Compounds
Organometallic Compounds
Salts and Inorganics
Solvents
Water
Lab Consumables
Products
Lab Consumables
Beakers and Lids
Bottles, Jars, and Jugs
Dishes
Dispensers
Filters and Filtration
Flasks
Microplates
Pipettes
Tubes
Vials
Lab Equipment and Instruments
Products
Lab Equipment and Instruments
Balances and Scales
Centrifuges
Cold Storage Products
Fume Hoods
Hotplates and Stirrers
Incubators
Microscopes
pH and Electrochemistry
Pumps
Shakers
Tubing and Accessories
Lab Furniture and Storage
Products
Lab Furniture and Storage
Desks and Tables
Fume Hoods
Laboratory Benches and Tables
Laboratory Carts and Accessories
Safety Cabinets
Seating
Shelving
Sinks and Drains
Storage Cabinets
Task Lighting
Life Sciences Products
Products
Life Sciences Products
Antibodies
Biochemical Reagents
Cell Analysis Products
Cell Culture Media
Cellular Imaging
Gel Electrophoresis Equipment and Supplies
Life Sciences Buffers
Molecular Biology Reagents and Kits
PCR Equipment and Supplies
Protein Analysis Reagents
Recombinant Proteins
Safety Products
Products
Safety Products
Chemical Detection
Cleaners and Disinfectants
Eye and Face Protection
Fall Protection and Confined Space Entry
Hand Protection
Lockout Tagout Safety
Radiation Monitoring Instrumentation
Respiratory Protection
Safety Clothing
Signs and Tags
Spill Control and Containment
Wipes and Wipe Accessories
Shop All Products
Browse Featured Brands
Applications
Cannabis
Cell Analysis Methods
Cell Culture
Chromatography
Controlled Environments
Genomics
Industrial Testing Applications
Laboratory Medicine
Microbiology Methods
Molecular Biology Methods
PCR/qPCR
Production Applications
Protein Biology Methods
Programs and Services
BioAdvantage
Cannabis Laboratory and Safety
Edge
Encompass Procurement Services
HealthPRO
New Lab Start-Up
Sustainability
Services
Programs and Services
Services
Safety Services
Thermo Fisher Financial Services
Unity Lab Services
Browse All Programs
Documents and Certificates
Main Menu
Documents and Certificates
Certificates
Safety Data Sheets
Special Offers
Main Menu
Special Offers
Flex Dollars
Earn Flex Dollars with any qualifying Thermo Scientific equipment purchase.
Spotlight
Spotlight showcases the latest and greatest products and our best special offers throughout the year.
All Special Offers
Take advantage of limited-time offers on everything from consumables to equipment.
View All Special Offers and Programs
Orders
Main Menu
Orders
My Lists
Quick Order
Order Status
Approve Requisitions
missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Home
Products
Personal Protective Equipment
Safety Clothing
Protective Coveralls
General Purpose Coveralls
Tians epic™ Polypro Coverall
19200883
Print
Tians epic™ Polypro Coverall
X-Large; 17L x 10W x 9 in.H
Supplier:
Tians 200881XL
View more versions of this product
Catalog No.
19-200-883
$100.06
/
Case of 25 Each
Quantity
Add to cart
Description
Specifications
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Size
X-Large
Color
White
Cuff Style
Elastic
SDS
Documents
Documents
Promotions
Promotions