missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tiagabine Hydrochloride, TCI America™

Manufacturer:  TCI America T316550MG

 View more versions of this product

Catalog No. T316550MG



Specifications

Specifications

Tiagabine Hydrochloride
193°C
145821-59-6
MFCD07369025
YUKARLAABCGMCN-PKLMIRHRSA-N
(3R)-1-[4,4-bis(3-methylthiophen-2-yl)but-3-enyl]piperidine-3-carboxylic acid;hydrochloride
91274
412.00
≥99.0% (HPLC)
White-Yellow
50mg
C20H26ClNO2S2
tiagabine hydrochloride, tiagabine hcl, gabitril, abbott 70569.hcl, tiagabine hydrochloride, unii-dqh6t6d8oy, dqh6t6d8oy, abbott-70569.hcl, r-1-4,4-bis 3-methylthiophen-2-yl but-3-en-1-yl piperidine-3-carboxylic acid hydrochloride, --r-1-4,4-bis 3-methyl-2-thienyl-3-butenyl nipecotic acid, hydrochloride
CC1=C(SC=C1)C(=CCCN2CCCC(C2)C(=O)O)C3=C(C=CS3)C.Cl
412.003
CHEBI:85388
Crystal-Powder at 20°C

Chemical Identifiers

145821-59-6
412.003
YUKARLAABCGMCN-PKLMIRHRSA-N
91274
(3R)-1-[4,4-bis(3-methylthiophen-2-yl)but-3-enyl]piperidine-3-carboxylic acid;hydrochloride
C20H26ClNO2S2
MFCD07369025
tiagabine hydrochloride, tiagabine hcl, gabitril, abbott 70569.hcl, tiagabine hydrochloride, unii-dqh6t6d8oy, dqh6t6d8oy, abbott-70569.hcl, r-1-4,4-bis 3-methylthiophen-2-yl but-3-en-1-yl piperidine-3-carboxylic acid hydrochloride, --r-1-4,4-bis 3-methyl-2-thienyl-3-butenyl nipecotic acid, hydrochloride
CHEBI:85388
CC1=C(SC=C1)C(=CCCN2CCCC(C2)C(=O)O)C3=C(C=CS3)C.Cl
Safety and Handling

Safety and Handling

RTECSNumber : TM6128245

TSCA : No

Recommended Storage : Refrigerator

Documents