Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thyroid Cancer Marker, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thyroid Cancer Marker, Mouse Monoclonal Antibody.

$268.31 - $1,158.43

Specifications

Antigen Thyroid Cancer Marker
Clone 373E1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Format Quantity Price Quantity    

MS1914R7

 
thermo scientific
MS1914R7
Prediluted 7mL Each for $818.88

MS1914S

 
thermo scientific™ lab vision™
MS1914S
Concentrated 1mL Each for $1,158.43

MS1914S0

 
thermo scientific™ lab vision™
MS1914S0
Concentrated 100µL Each for $268.31

MS1914S1

 
thermo scientific™ lab vision™
MS1914S1
Concentrated 500µL Each for $760.47
Description & Specifications

Specifications

Antigen Thyroid Cancer Marker
Clone 373E1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Purification Method Papillary Thyroid CA

Provide accurate, reproducible results with the Thermo Scientific™ Thyroid Cancer Marker, Mouse Monoclonal Antibody.

Papillary carcinoma is by far the most frequent malignant tumor of the thyroid with a threefold prevelance in females. Studies have shown that 100% of malignant thyroid lesions were found to contain the keratan sulfate (KS) bearing molecules, while these were virtually absent from benign tissues and other thyroid tumors, with the exception of 21% of follicular carcinoma cases analyzed. Clone 373E1 is specific for the keratan sulfate bearing molecular complex.

Host Species: Mouse

Clone: 373E1

Species Reactivity: Human. Others not tested.

Positive Control: Papillary Thyroid CA

Cellular Localization: Membrane, cytoplasmic and luminal

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO