Please login to your online account to display your discounted pricing

Alfa Aesar™ Thymopoietin II Peptide Fragment

For cellular and molecular biology applications

Specifications

Product Type Thymopoietin II Peptide Fragment
Molecular Weight 1610.83
Storage Requirements -30° to -10°C
Sequence Gly-Glu-Gln-Arg-Lys-Asp-Val-Tyr-Val-Gln-Leu-Tyr-Leu
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66498EXD

 
alfa aesar™
J66498EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66498MCR

 
alfa aesar™
J66498MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Thymopoietin II Peptide Fragment
Molecular Weight 1610.83
Storage Requirements -30° to -10°C
Sequence Gly-Glu-Gln-Arg-Lys-Asp-Val-Tyr-Val-Gln-Leu-Tyr-Leu
Format Solid
Regulatory Status RUO
Purification Method HPLC