Please login to your online account to display your discounted pricing

Alfa Aesar™ Thymopoietin II (32-35)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66664LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66664LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Sequence Arg-Lys-Asp-Val
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Thymopoietin II (32-35)
Molecular Weight 516.60
Purification Method HPLC