Please login to your online account to display your discounted pricing

Alfa Aesar™ Thymopoietin II (32-34)

For cellular and molecular biology applications

Specifications

Product Type Thymopoietin II (32-34)
Molecular Weight 417.50
Storage Requirements -30° to -10°C
Sequence Arg-Lys-Asp
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66634LB0

 
alfa aesar™
J66634LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66634MC

 
alfa aesar™
J66634MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Thymopoietin II (32-34)
Molecular Weight 417.50
Storage Requirements -30° to -10°C
Sequence Arg-Lys-Asp
Format Solid
Common Name TP-3, Thymotrinan
Regulatory Status RUO
Purification Method HPLC