Please login to your online account to display your discounted pricing

Alfa Aesar™ Thymopoietin II (32-34)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66634MC

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66634MC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Product Type Thymopoietin II (32-34)
Regulatory Status RUO
Format Solid
Purification Method HPLC
Molecular Weight 417.50
Quantity 100mg
Sequence Arg-Lys-Asp
Common Name TP-3, Thymotrinan