missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thymolphthalein, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  J.T.Baker V85702

Catalog No. 02004482



Specifications

Specifications

Thymolphthalein
10g
C28H30O4
LDKDGDIWEUUXSH-UHFFFAOYSA-N
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
31316
BAKER ANALYZED™ ACS Reagent
Wide Mouth Amber Glass Bottle
125-20-2
1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 5',5-diisopropyl-2',2-dimethylphenolphthalein, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
430.544
430.55

Chemical Identifiers

125-20-2
430.544
1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 5',5-diisopropyl-2',2-dimethylphenolphthalein, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
C28H30O4
LDKDGDIWEUUXSH-UHFFFAOYSA-N
31316
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
Documents