Please login to your online account to display your discounted pricing

Thymolphthalein, ACS Grade, LabChem™

$54.80 - $54.80

Chemical Identifiers

Chemical Name or Material Thymolphthalein
CAS 125-20-2
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C28H30O4
Formula Weight 430.5
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC260607

 
lab chem
LC260607
Amber Glass 5g Each for $54.80
Description & Specifications

Specifications

Formula Weight 430.5
Quantity 5g
CAS Max % 100
Color White
Packaging Amber Glass
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS 125-20-2
Health Hazard 2 GHS H Statement
Product is not hazardous.
Linear Formula C28H30O4
Chemical Name or Material Thymolphthalein
Purity Grade ACS Grade
Physical Form Powder
Decomposition Information Carbon monoxide; Carbon dioxide
Storage Information Room Temperature
Identification Passes Test
Safety and Handling

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Product is not hazardous.