Please login to your online account to display your discounted pricing

Thymolphthalein, 0.1% in Ethanol, Certified, LabChem™

$54.40 - $54.40

Chemical Identifiers

Chemical Name or Material Thymolphthalein
CAS 64-17-5
CAS Max % 90.4
CAS 67-63-0
CAS Max % 5
CAS 67-56-1
CAS Max % 4.5
CAS 125-20-2
CAS Max % 0.1
Name Note 0.1% in Ethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC260757

 
lab chem
LC260757
Poly Bottle 125mL Each for $54.40
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 430.5
Chemical Name or Material Thymolphthalein
CAS Max % 4.5
Identification Passes Test
CAS 125-20-2
CAS Max % 0.1
Name Note 0.1% in Ethanol
Physical Form Liquid
Linear Formula C28H30O4
Packaging Poly Bottle
CAS 64-17-5
UN Number UN1170
Solubility Soluble in water
Health Hazard 1 Danger
Storage Information Room Temperature
CAS Max % 90.4
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes skin and serious eye irritation.
May cause respiratory irritation.
Causes damage to organs (central nervous system, optic nerve).
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
CAS 67-63-0
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Color Clear, colorless
Quantity 125mL
CAS Max % 5
CAS 67-56-1
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Highly flammable liquid and vapor.
Causes skin and serious eye irritation.
May cause respiratory irritation.
Causes damage to organs (central nervous system, optic nerve).