Please login to your online account to display your discounted pricing

Thymolphthalein, 0.05% w/v solution in ethanol, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Thymolphthalein
Name Note 0.05% w/v solution in ethanol
CAS 125-20-2
Physical Form Liquid
Molecular Formula C28H30O4
Formula Weight 430.55
MDL Number MFCD00005909
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38706AP

  SDS
alfa aesar
38706AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 430.55
Quantity 500mL
Color Colorless
DOT Information DOT Class: 3, Packaging Group: II
Physical Form Liquid
Density 0.92g/cm3 at 20°C
Name Note 0.05% w/v solution in ethanol
Health Hazard 1 H225-H371-H313-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful in contact with skin. May be harmful if inhaled.
CAS 125-20-2
Appearance Colorless liquid
Vapor Pressure 57 hPa (150mm Hg) at 20°C
MDL Number MFCD00005909
Chemical Name or Material Thymolphthalein
Molecular Formula C28H30O4