Thymolphthalein, 0.05% in Ethanol, Certified, LabChem™
Manufacturer:  Lab Chem LC260701
 View more versions of this product
Catalog No. LC260701
Specifications
Specifications
Thymolphthalein
90.4,5,4.55,0.05
C28H30O4
430.5
Certified
Soluble in water
Blue to Colorless
500mL
0.05% in Ethanol
67-63-0,67-56-1,125-20-2
Passes Test
Liquid
Poly Bottle
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
9.3 to 10.5
Documents