Please login to your online account to display your discounted pricing

Thymol Blue Sodium Salt, For analysis ACS, ACROS Organics™

$70.87 - $441.30

Chemical Identifiers

Chemical Name or Material Thymol Blue sodium salt
CAS 62625-21-2
Purity Grade ACS Reagent Grade
Molecular Formula C27H29NaO5S
Formula Weight 488.58
MDL Number MFCD00151093
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC420941000

  SDS
thermo fisher scientific
AC420941000
Glass bottle 100g Each for $441.30

AC420940100

  SDS
thermo fisher scientific
420940100
Glass bottle 10g Each for $70.87

AC420940250

  SDS
thermo fisher scientific
420940250
Glass bottle 25g Each for $120.91
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 488.58
Purity Grade ACS Reagent Grade
Quantity 100g
Color Dark orange to green-black or brown-black
Molecular Formula C27H29NaO5S
CAS 62625-21-2
Packaging Glass bottle
Clarity Passes Test
MDL Number MFCD00151093
Infrared Spectrum Authentic
Visual Transition Interval Acid range: from pH 1.2 (pink) to pH 2.8 (yellow) Alkaline range: from pH 8.0 (yellow) to pH 9.2 (blue)
Chemical Name or Material Thymol Blue sodium salt