missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thymol Blue Sodium Salt, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RDCT04205F1
Specifications
Thymol Blue Sodium Salt | |
98.0 | |
C27H29NaO5S | |
OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate | |
23692293 | |
Pouch |
62625-21-2 | |
100.0 | |
MFCD00151093 | |
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O | |
488.57 | |
ACS | |
5 g |
Chemical Identifiers
62625-21-2 | |
488.57 | |
OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate |
C27H29NaO5S | |
MFCD00151093 | |
23692293 | |
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Safety and Handling
ShelfLife : 60 months